Product Details

SNP ID
rs73045078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41681674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAATTCTGATACATACTACAAC[A/G]TGAATGAACCTTAAAGACGTTATGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEACAM7 PubMed Links

Gene Details

Gene
CEACAM7
Gene Name
carcinoembryonic antigen related cell adhesion molecule 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291485.1 Intron NP_001278414.1
NM_006890.4 Intron NP_008821.2

View Full Product Details