Product Details

SNP ID
rs73042059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32582097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTGGGGAGTTGTTTCTACCAC[C/T]GCAAGAGACTGTTATGTATAGGAAA
Phenotype
MIM: 604583
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDCD5 PubMed Links

Gene Details

Gene
PDCD5
Gene Name
programmed cell death 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004708.3 Intron NP_004699.1
XM_005259392.4 Intron XP_005259449.1

View Full Product Details