Product Details

SNP ID
rs72625387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:2961403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACATTACTTCAACCTCAATGTCT[A/G]TGCCAGAACATGTTTTAACGAGTGA
Phenotype
MIM: 490000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFY PubMed Links

Gene Details

Gene
ZFY
Gene Name
zinc finger protein, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145275.1 591 Missense Mutation ATG,GTG M105V NP_001138747.1
NM_001145276.1 591 Intron NP_001138748.1
NM_003411.3 591 Missense Mutation ATG,GTG M131V NP_003402.2
XM_005262570.2 591 Missense Mutation ATG,GTG M131V XP_005262627.1
XM_011531473.2 591 Missense Mutation ATG,GTG M105V XP_011529775.1
XM_011531474.2 591 Missense Mutation ATG,GTG M105V XP_011529776.1
XM_011531475.2 591 Missense Mutation ATG,GTG M131V XP_011529777.1
XM_017030075.1 591 Missense Mutation ATG,GTG M131V XP_016885564.1
XM_017030076.1 591 Missense Mutation ATG,GTG M131V XP_016885565.1
XM_017030077.1 591 Missense Mutation ATG,GTG M105V XP_016885566.1

View Full Product Details