Product Details
- SNP ID
-
rs72680254
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:124668171 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATACCTATTTTCCCACTAACATC[A/C]CCTATATTATTAATGAAGTATCCTT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD50
PubMed Links
Gene Details
- Gene
- ANKRD50
- Gene Name
- ankyrin repeat domain 50
View Full Product Details