Product Details

SNP ID
rs74113801
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:109250506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCCGTGCTTAAAGGCTCCACGG[C/T]TCAGATGCCAGTCCTGCAAGCTGGC
Phenotype
MIM: 604265
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELSR2 PubMed Links

Gene Details

Gene
CELSR2
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001408.2 488 Missense Mutation CTC,TTC L143F NP_001399.1
XM_005270580.4 488 Missense Mutation CTC,TTC L143F XP_005270637.1

View Full Product Details