Product Details
- SNP ID
-
rs73667677
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:135665817 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGTGGGGATGGGAGGTGTGCCTG[C/T]GGGGTCCCTGTCCCTGCGCTGAGAG
- Phenotype
-
MIM: 612903
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LCN9
PubMed Links
Gene Details
- Gene
- LCN9
- Gene Name
- lipocalin 9
View Full Product Details