Product Details

SNP ID
rs71454740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108591558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGACACTGGGGTGGACACTGCTGC[A/C]AGCACAGGGGCTTTCGGGGGGACCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM119 PubMed Links

Gene Details

Gene
TMEM119
Gene Name
transmembrane protein 119
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181724.2 1179 Missense Mutation GGC,TGC G276C NP_859075.2
XM_011538271.2 1179 Missense Mutation GGC,TGC G276C XP_011536573.1

View Full Product Details