Product Details
- SNP ID
-
rs72860147
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:138669987 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAATCCATAATCCTGAAATTTCCT[C/T]CCTCTACTTCCCACTTTTGTTTTCT
- Phenotype
-
MIM: 604635
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NXPH2
PubMed Links
Gene Details
- Gene
- NXPH2
- Gene Name
- neurexophilin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007226.2 |
1730 |
UTR 3 |
|
|
NP_009157.1 |
View Full Product Details