Product Details
- SNP ID
-
rs72613822
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:25380626 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACAAGGAGCTCTAAGGGCAAGAG[A/G]CACAGAAAGACAGAAGAGATGCAGA
- Phenotype
-
MIM: 602415
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DTNB
PubMed Links
Gene Details
- Gene
- DTNB
- Gene Name
- dystrobrevin beta
View Full Product Details