Product Details

SNP ID
rs73958874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:57147611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAAACGACAACCAACTGCACG[A/G]TGGTCTGAGGCAGAGTGTGGTTCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BOD1L2 PubMed Links

Gene Details

Gene
BOD1L2
Gene Name
biorientation of chromosomes in cell division 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257964.1 550 Missense Mutation GAT,GGT D100G NP_001244893.1

View Full Product Details