Product Details

SNP ID
rs73586077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:146548049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATATAGTCTTTCTGCAGAATAG[C/T]GATTTTGAACAGTTGGCATATCATG
Phenotype
MIM: 612906
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB32 PubMed Links

Gene Details

Gene
RAB32
Gene Name
RAB32, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006834.4 Intron NP_006825.1

View Full Product Details