Product Details

SNP ID
rs73617919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149447650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGTGGTAAAAAATGATTTATTT[C/T]TGAAGCATGGTTAACAGCTATGCAA
Phenotype
MIM: 611106
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZC3H12D PubMed Links

Gene Details

Gene
ZC3H12D
Gene Name
zinc finger CCCH-type containing 12D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207360.2 4887 UTR 3 NP_997243.2

View Full Product Details