Product Details

SNP ID
rs72995576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149825164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGTAAAAAGAAAGTTTTCTCAA[C/T]GTTTATAACTTAAGCTTATATTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP11 PubMed Links

Gene Details

Gene
LRP11
Gene Name
LDL receptor related protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032832.5 Intron NP_116221.3
XM_011536194.2 Intron XP_011534496.1
XM_017011380.1 Intron XP_016866869.1

View Full Product Details