Product Details

SNP ID
rs68001052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:85533142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAGAAAAGTAGGAGGGACTCCG[A/G]GAGAGAGGGAGGGAATGAGGGAGGG
Phenotype
MIM: 604807
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLRT2 PubMed Links

Gene Details

Gene
FLRT2
Gene Name
fibronectin leucine rich transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013231.4 Intron NP_037363.1
XM_005267490.3 Intron XP_005267547.1
XM_011536611.2 Intron XP_011534913.1
XM_017021129.1 Intron XP_016876618.1
XM_017021130.1 Intron XP_016876619.1
XM_017021131.1 Intron XP_016876620.1
XM_017021132.1 Intron XP_016876621.1
XM_017021133.1 Intron XP_016876622.1
XM_017021134.1 Intron XP_016876623.1
XM_017021135.1 Intron XP_016876624.1
Gene
LOC100506731
Gene Name
uncharacterized LOC100506731
There are no transcripts associated with this gene.

View Full Product Details