Product Details

SNP ID
rs74090094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50176315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATTATGAGAAGCACAGATATCA[C/T]CAGAAAAGAAAGCAATCATTTGGAG
Phenotype
MIM: 615335 MIM: 608364
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CERS5 PubMed Links

Gene Details

Gene
CERS5
Gene Name
ceramide synthase 5
There are no transcripts associated with this gene.

Gene
LIMA1
Gene Name
LIM domain and actin binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113546.1 3087 UTR 3 NP_001107018.1
NM_001113547.1 3087 UTR 3 NP_001107019.1
NM_001243775.1 3087 UTR 3 NP_001230704.1
NM_016357.4 3087 UTR 3 NP_057441.1
XM_011538455.2 3087 UTR 3 XP_011536757.1
XM_017019429.1 3087 UTR 3 XP_016874918.1
XM_017019430.1 3087 UTR 3 XP_016874919.1

View Full Product Details