Product Details

SNP ID
rs71486131
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97713972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTTCCTGCGCAGCCCGCTGGG[C/G]GTGTTGCGGCTGCTGCAGCTGCTGG
Phenotype
MIM: 616970
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
MARVELD1 PubMed Links

Gene Details

Gene
MARVELD1
Gene Name
MARVEL domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031484.3 265 Silent Mutation GGC,GGG G32G NP_113672.1

View Full Product Details