Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001323573.1 | Intron | NP_001310502.1 | |||
NM_001323574.1 | Intron | NP_001310503.1 | |||
NM_001323575.1 | Intron | NP_001310504.1 | |||
NM_032785.3 | Intron | NP_116174.3 | |||
XM_011542308.2 | Intron | XP_011540610.1 | |||
XM_011542310.2 | Intron | XP_011540612.1 | |||
XM_017002595.1 | Intron | XP_016858084.1 | |||
XM_017002596.1 | Intron | XP_016858085.1 | |||
XM_017002597.1 | Intron | XP_016858086.1 | |||
XM_017002598.1 | Intron | XP_016858087.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001302082.1 | Intron | NP_001289011.1 | |||
NM_024603.3 | Intron | NP_078879.2 | |||
XM_011542141.2 | Intron | XP_011540443.1 | |||
XM_011542142.2 | Intron | XP_011540444.1 | |||
XM_017002331.1 | Intron | XP_016857820.1 | |||
XM_017002332.1 | Intron | XP_016857821.1 | |||
XM_017002333.1 | Intron | XP_016857822.1 | |||
XM_017002334.1 | Intron | XP_016857823.1 |