Product Details

SNP ID
rs74082414
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:53068096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACTGAGCACTTACTATGTGCAG[C/T]ACATGCATTTCACTGACCCCTCTCA
Phenotype
MIM: 608661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PODN PubMed Links

Gene Details

Gene
PODN
Gene Name
podocan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153703.4 Intron NP_714914.2
XM_005270457.3 Intron XP_005270514.1
XM_011540677.2 Intron XP_011538979.1
XM_017000294.1 Intron XP_016855783.1

View Full Product Details