Product Details

SNP ID
rs74064168
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:32681143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAATCGCTTAGTTTTTTGTTGTT[A/G]TTGTTTTTTAACTGAGTAGATCATT
Phenotype
MIM: 602923 MIM: 611750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBBP4 PubMed Links

Gene Details

Gene
RBBP4
Gene Name
RB binding protein 4, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135255.1 2775 UTR 3 NP_001128727.1
NM_001135256.1 2775 UTR 3 NP_001128728.1
NM_005610.2 2775 UTR 3 NP_005601.1
Gene
SYNC
Gene Name
syncoilin, intermediate filament protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161708.1 2775 UTR 3 NP_001155180.1
NM_030786.2 2775 UTR 3 NP_110413.2

View Full Product Details