Product Details

SNP ID
rs66501904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:106684694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAAGAGCTATAGGTGCCAAATT[A/G]TATTTTAAAGTATCCCTTGCAGAGT
Phenotype
MIM: 601244
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GUCY1A2 PubMed Links

Gene Details

Gene
GUCY1A2
Gene Name
guanylate cyclase 1 soluble subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000855.2 5444 UTR 3 NP_000846.1
NM_001256424.1 5444 UTR 3 NP_001243353.1

View Full Product Details