Product Details

SNP ID
rs72792760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:133981365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTATCTGCCATGAAGGCAGCTA[C/T]CTAAATAGCAGCTTTTAAAGAGACG
Phenotype
MIM: 604492
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VDAC1 PubMed Links

Gene Details

Gene
VDAC1
Gene Name
voltage dependent anion channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003374.2 Intron NP_003365.1
XM_005272075.3 Intron XP_005272132.1
XM_017009821.1 Intron XP_016865310.1
XM_017009822.1 Intron XP_016865311.1
XM_017009823.1 Intron XP_016865312.1

View Full Product Details