Product Details

SNP ID
rs79425787
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:1234257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGTGGTCTTTGTTGTCCCACTT[C/T]GGGACCGGGTCCAGTCTGGGGTCTA
Phenotype
MIM: 615291 MIM: 616593 MIM: 614282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GALT6 PubMed Links

Gene Details

Gene
B3GALT6
Gene Name
beta-1,3-galactosyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080605.3 2009 UTR 3 NP_542172.2
Gene
FAM132A
Gene Name
family with sequence similarity 132 member A
There are no transcripts associated with this gene.

Gene
SDF4
Gene Name
stromal cell derived factor 4
There are no transcripts associated with this gene.

View Full Product Details