Product Details
- SNP ID
-
rs74511182
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:1242864 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGCAGCAGCCCCTGCACCTGTA[C/G]CGTGAAGACCCTGCTGTTGCTCTCC
- Phenotype
-
MIM: 615291
MIM: 616593
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
B3GALT6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11260566] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- B3GALT6
- Gene Name
- beta-1,3-galactosyltransferase 6
There are no transcripts associated with this gene.
- Gene
- FAM132A
- Gene Name
- family with sequence similarity 132 member A
View Full Product Details