Product Details

SNP ID
rs75855672
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:158610638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATTTTGAAGTATGGAAAAAAA[A/C]CAAGAAGAAAACAGGTGTTATGCTG
Phenotype
MIM: 182860
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR10Z1 PubMed Links

Gene Details

Gene
OR10Z1
Gene Name
olfactory receptor family 10 subfamily Z member 1
There are no transcripts associated with this gene.

Gene
SPTA1
Gene Name
spectrin alpha, erythrocytic 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003126.2 7981 Intron NP_003117.2
XM_011509916.2 7981 UTR 3 XP_011508218.1
XM_011509917.2 7981 UTR 3 XP_011508219.1
XM_011509918.2 7981 Intron XP_011508220.1
XM_011509919.2 7981 Intron XP_011508221.1

View Full Product Details