Product Details

SNP ID
rs78598639
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:158612883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGCTTGGAAGGCATTCTCTAT[G/T]TCATCACTGGACTTGATGTTTTCTG
Phenotype
MIM: 182860
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10Z1 PubMed Links

Gene Details

Gene
OR10Z1
Gene Name
olfactory receptor family 10 subfamily Z member 1
There are no transcripts associated with this gene.

Gene
SPTA1
Gene Name
spectrin alpha, erythrocytic 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003126.2 7163 Missense Mutation GAA,GAC E2356D NP_003117.2
XM_011509916.2 7163 Missense Mutation GAA,GAC E2356D XP_011508218.1
XM_011509917.2 7163 Missense Mutation GAA,GAC E2350D XP_011508219.1
XM_011509918.2 7163 Intron XP_011508220.1
XM_011509919.2 7163 Intron XP_011508221.1

View Full Product Details