Product Details

SNP ID
rs78637010
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:156644729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTCAGGCTTAGGAAAGTGCAGG[A/G]ACAGGTATGAAGGCAGATGCTGAGA
Phenotype
MIM: 600347
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCAN PubMed Links

Gene Details

Gene
BCAN
Gene Name
brevican
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021948.4 Intron NP_068767.3
NM_198427.1 Intron NP_940819.1
XM_011509866.1 Intron XP_011508168.1
XM_017002047.1 Intron XP_016857536.1

View Full Product Details