Product Details

SNP ID
rs80173211
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:26282538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTAGAGGGTGGGCGGGAATGTGC[C/T]GAAGATCTCAAAGGCAGAGGCATCC
Phenotype
MIM: 615679 MIM: 609151
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP85 PubMed Links

Gene Details

Gene
CEP85
Gene Name
centrosomal protein 85
There are no transcripts associated with this gene.

Gene
SH3BGRL3
Gene Name
SH3 domain binding glutamate rich protein like 3
There are no transcripts associated with this gene.

Gene
UBXN11
Gene Name
UBX domain protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077262.1 1112 Missense Mutation AGC,GGC S322G NP_001070730.1
NM_145345.2 1112 Missense Mutation AGC,GGC S409G NP_663320.2
NM_183008.2 1112 Missense Mutation AGC,GGC S442G NP_892120.2

View Full Product Details