Product Details
- SNP ID
-
rs78295934
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:206938588 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTTTTAACTGCATTCTCTTTACT[C/T]GAAACTGTAACATTTGTTTGTATAC
- Phenotype
-
MIM: 609563
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPO
PubMed Links
Gene Details
- Gene
- CPO
- Gene Name
- carboxypeptidase O
View Full Product Details