Product Details

SNP ID
rs75906230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207822315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATAAAAAGAAAACCTTTGAAAG[G/T]TGAAAGATAAGAAAATCCTTGTGTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLEKHM3 PubMed Links

Gene Details

Gene
PLEKHM3
Gene Name
pleckstrin homology domain containing M3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080475.2 8780 UTR 3 NP_001073944.1
XM_005246545.3 8780 UTR 3 XP_005246602.1
XM_006712521.3 8780 Intron XP_006712584.1
XM_011511160.2 8780 UTR 3 XP_011509462.1
XM_011511163.2 8780 Intron XP_011509465.1
XM_017004071.1 8780 Intron XP_016859560.1
XM_017004072.1 8780 UTR 3 XP_016859561.1
XM_017004073.1 8780 UTR 3 XP_016859562.1

View Full Product Details