Product Details

SNP ID
rs80265687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219600688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATAACAATGCCTTAGTGCTATAA[G/T]GAATAGTTATTCTATGACATACTTT
Phenotype
MIM: 607172
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
STK11IP PubMed Links

Gene Details

Gene
STK11IP
Gene Name
serine/threonine kinase 11 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052902.3 Intron NP_443134.3
XM_011510526.1 Intron XP_011508828.1
XM_011510527.2 Intron XP_011508829.1
XM_011510528.2 Intron XP_011508830.1
XM_011510529.1 Intron XP_011508831.1

View Full Product Details