Product Details

SNP ID
rs730905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28713305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACTTCAAAGAGTCTCTGTTTGG[A/C]CAGTAAATTATTTGGTGACCTCATG
Phenotype
MIM: 605744
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EXTL3 PubMed Links

Gene Details

Gene
EXTL3
Gene Name
exostosin like glycosyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001440.3 Intron NP_001431.1
XM_011544440.2 Intron XP_011542742.1

View Full Product Details