Product Details

SNP ID
rs338078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:44943296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCATCTGGTTCTTATTTCTGTGC[A/G]TTGCAAAACCCCGAGAGAAATCCAC
Phenotype
MIM: 603714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIX3 PubMed Links

Gene Details

Gene
SIX3
Gene Name
SIX homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005413.3 Intron NP_005404.1
Gene
SIX3-AS1
Gene Name
SIX3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details