Product Details

SNP ID
rs1543914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56385818 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCTTAAGGCAGCCAATTACAGCT[A/C]TTCCATAGTTTTTGTTTAGAAAAAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SDR16C6P PubMed Links

Gene Details

Gene
SDR16C6P
Gene Name
short chain dehydrogenase/reductase family 16C, member 6, pseudogene
There are no transcripts associated with this gene.

View Full Product Details