Product Details
- SNP ID
-
rs168726
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:180849402 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ACATTCATCACATGTATGGGGTGTC[C/T]TGCCAGGGTGGGTACTCTTATGGTT
- Phenotype
-
MIM: 610281
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZFP62
PubMed Links
Gene Details
- Gene
- ZFP62
- Gene Name
- ZFP62 zinc finger protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001172638.1 |
2263 |
Missense Mutation |
AAG,AGG |
K698R |
NP_001166109.1 |
NM_152283.4 |
2263 |
Missense Mutation |
AAG,AGG |
K665R |
NP_689496.4 |
XM_017009710.1 |
2263 |
Missense Mutation |
AAG,AGG |
K665R |
XP_016865199.1 |
XM_017009711.1 |
2263 |
Missense Mutation |
AAG,AGG |
K665R |
XP_016865200.1 |
XM_017009712.1 |
2263 |
Missense Mutation |
AAG,AGG |
K665R |
XP_016865201.1 |
XM_017009713.1 |
2263 |
Missense Mutation |
AAG,AGG |
K665R |
XP_016865202.1 |
XM_017009714.1 |
2263 |
Missense Mutation |
AAG,AGG |
K665R |
XP_016865203.1 |
XM_017009715.1 |
2263 |
Missense Mutation |
AAG,AGG |
K665R |
XP_016865204.1 |
XM_017009716.1 |
2263 |
Missense Mutation |
AAG,AGG |
K715R |
XP_016865205.1 |
XM_017009717.1 |
2263 |
Missense Mutation |
AAG,AGG |
K698R |
XP_016865206.1 |
XM_017009718.1 |
2263 |
Missense Mutation |
AAG,AGG |
K715R |
XP_016865207.1 |
XM_017009719.1 |
2263 |
Missense Mutation |
AAG,AGG |
K715R |
XP_016865208.1 |
XM_017009720.1 |
2263 |
Missense Mutation |
AAG,AGG |
K698R |
XP_016865209.1 |
XM_017009721.1 |
2263 |
Missense Mutation |
AAG,AGG |
K665R |
XP_016865210.1 |
View Full Product Details