Product Details
- SNP ID
-
rs11896926
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:168872431 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAATGAGACTCAAAGTCACGAAT[A/C]ACAGTATTGTCCTTGTATGTAGTTG
- Phenotype
-
MIM: 607496
MIM: 609395
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NOSTRIN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112521678] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NOSTRIN
- Gene Name
- nitric oxide synthase trafficking
There are no transcripts associated with this gene.
- Gene
- SPC25
- Gene Name
- SPC25, NDC80 kinetochore complex component
View Full Product Details