Product Details

SNP ID
rs545111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27637749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAGAGAGCAGTGCCCACTTCAT[G/T]TAGGCAAAGGGTGGAAGTGGGGAGA
Phenotype
MIM: 602728
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SCARA3 PubMed Links

Gene Details

Gene
SCARA3
Gene Name
scavenger receptor class A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016240.2 Intron NP_057324.2
NM_182826.1 Intron NP_878185.1
XM_017013535.1 Intron XP_016869024.1
XM_017013536.1 Intron XP_016869025.1
XM_017013537.1 Intron XP_016869026.1

View Full Product Details