Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001162861.2 | 2088 | Intron | NP_001156333.1 | ||
NM_001162862.2 | 2088 | Intron | NP_001156334.1 | ||
NM_001321518.1 | 2088 | Intron | NP_001308447.1 | ||
NM_004375.4 | 2088 | Intron | NP_004366.1 | ||
XM_011524342.2 | 2088 | UTR 5 | XP_011522644.1 | ||
XM_017024192.1 | 2088 | UTR 5 | XP_016879681.1 | ||
XM_017024193.1 | 2088 | Intron | XP_016879682.1 | ||
XM_017024194.1 | 2088 | Intron | XP_016879683.1 | ||
XM_017024195.1 | 2088 | Intron | XP_016879684.1 | ||
XM_017024196.1 | 2088 | Intron | XP_016879685.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_178509.5 | 2088 | Intron | NP_848604.3 | ||
XM_005257187.4 | 2088 | Intron | XP_005257244.1 | ||
XM_006721797.3 | 2088 | Intron | XP_006721860.1 | ||
XM_006721798.3 | 2088 | Intron | XP_006721861.1 | ||
XM_017024410.1 | 2088 | Intron | XP_016879899.1 | ||
XM_017024411.1 | 2088 | Intron | XP_016879900.1 | ||
XM_017024412.1 | 2088 | Intron | XP_016879901.1 | ||
XM_017024413.1 | 2088 | Intron | XP_016879902.1 | ||
XM_017024414.1 | 2088 | Intron | XP_016879903.1 | ||
XM_017024415.1 | 2088 | Intron | XP_016879904.1 |