Product Details

SNP ID
rs11633872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90537280 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTACATTTGACATACCACTCTTGT[A/G]TCTGTGTCATTTAATCCTCACTTTA
Phenotype
MIM: 608986
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRTC3 PubMed Links

Gene Details

Gene
CRTC3
Gene Name
CREB regulated transcription coactivator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042574.2 Intron NP_001036039.1
NM_022769.4 Intron NP_073606.3
XM_005254968.2 Intron XP_005255025.1
XM_011521906.2 Intron XP_011520208.1

View Full Product Details