Product Details

SNP ID
rs1790761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67454383 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTAGTTTGGTGCCCTCCGTACCC[C/T]CAGGGTTCCTCCTCAGGCTGACCCC
Phenotype
MIM: 608965
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABP4 PubMed Links

Gene Details

Gene
CABP4
Gene Name
calcium binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300895.1 Intron NP_001287824.1
NM_001300896.1 Intron NP_001287825.1
NM_145200.3 Intron NP_660201.1
XM_005274114.3 Intron XP_005274171.2
XM_011545181.2 Intron XP_011543483.1
XM_011545182.2 Intron XP_011543484.1
XM_011545183.2 Intron XP_011543485.1
XM_017018025.1 Intron XP_016873514.1
Gene
GPR152
Gene Name
G protein-coupled receptor 152
There are no transcripts associated with this gene.

Gene
TMEM134
Gene Name
transmembrane protein 134
There are no transcripts associated with this gene.

View Full Product Details