Product Details

SNP ID
rs3810990
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:6102585 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GTACCCGAGCCCACTCTCCACTCCA[C/T]CTCCTTGGGCCTCAAGTGTCCTGGG
Phenotype
MIM: 610771 MIM: 601142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHD5 PubMed Links

Gene Details

Gene
CHD5
Gene Name
chromodomain helicase DNA binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015557.2 8865 UTR 3 NP_056372.1
Gene
KCNAB2
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 2
There are no transcripts associated with this gene.

View Full Product Details