Product Details

SNP ID
rs753213
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:17962849 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACACTGCAGACTGCAGGGCATTTTC[A/T]GGCTACATAGCCTCAAGTTCCTAGA
Phenotype
MIM: 615076 MIM: 605937
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MGME1 PubMed Links

Gene Details

Gene
MGME1
Gene Name
mitochondrial genome maintenance exonuclease 1
There are no transcripts associated with this gene.

Gene
SNORD17
Gene Name
small nucleolar RNA, C/D box 17
There are no transcripts associated with this gene.

Gene
SNX5
Gene Name
sorting nexin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282454.1 Intron NP_001269383.1
NM_014426.3 Intron NP_055241.1
NM_152227.2 Intron NP_689413.1

View Full Product Details