Product Details

SNP ID
rs9425343
Assay Type
Functionally tested
NCBI dbSNP Submissions
83
Location
Chr.1:184694403 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCAACCTCCTGAGAACTTGATGAA[A/C]TCTGTTCACCACTCTGATTCTGAGA
Phenotype
MIM: 610214
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EDEM3 PubMed Links

Gene Details

Gene
EDEM3
Gene Name
ER degradation enhancing alpha-mannosidase like protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319960.1 2720 Missense Mutation AGT,ATT S836I NP_001306889.1
NM_025191.3 2720 Missense Mutation AGT,ATT S820I NP_079467.3
XM_005245499.1 2720 Missense Mutation AGT,ATT S821I XP_005245556.1
XM_011510012.1 2720 Missense Mutation AGT,ATT S744I XP_011508314.1
XM_011510013.2 2720 Intron XP_011508315.1
XM_011510014.2 2720 Intron XP_011508316.1
XM_017002397.1 2720 Missense Mutation AGT,ATT S743I XP_016857886.1
XM_017002398.1 2720 Intron XP_016857887.1

View Full Product Details