Product Details

SNP ID
rs1448938
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:30871277 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGAAAATTCACATGAGGACCAAGC[A/G]CAGAGAAACAGGGAAGTCAAGAGCA
Phenotype
MIM: 612321
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCDC5 PubMed Links

Gene Details

Gene
DCDC5
Gene Name
doublecortin domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020869.3 Intron NP_065920.2

View Full Product Details