Product Details

SNP ID
rs269970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:45197007 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGACTTGGACTAATGTTGGGATCA[C/T]TACGAAGGATGTGCTGGCTTCAGAA
Phenotype
MIM: 601611
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC14A2 PubMed Links

Gene Details

Gene
SLC14A2
Gene Name
solute carrier family 14 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242692.1 Intron NP_001229621.1
NM_007163.3 Intron NP_009094.3
XM_017026015.1 Intron XP_016881504.1
XM_017026016.1 Intron XP_016881505.1

View Full Product Details