Product Details

SNP ID
rs9830360
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:13558113 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTGGGCACAGCAAAGGCCTGAGGGA[C/G]TCGCAGAGAGACTGGGCTGCCCGCT
Phenotype
MIM: 135821
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBLN2 PubMed Links

Gene Details

Gene
FBLN2
Gene Name
fibulin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004019.1 Intron NP_001004019.1
NM_001165035.1 Intron NP_001158507.1
NM_001998.2 Intron NP_001989.2
XM_006713026.2 Intron XP_006713089.1

View Full Product Details