Product Details

SNP ID
rs10758307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35501911 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGTCAAGATATTTTTTCTAGCC[A/G]TGCTTTCTAGTTTATAATATGAAAA
Phenotype
MIM: 611053
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL36AP33 PubMed Links

Gene Details

Gene
RPL36AP33
Gene Name
ribosomal protein L36a pseudogene 33
There are no transcripts associated with this gene.

Gene
RUSC2
Gene Name
RUN and SH3 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135999.1 Intron NP_001129471.1
XM_006716896.2 Intron XP_006716959.1

View Full Product Details