Product Details

SNP ID
rs2242029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:129055225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTCCCCGCCCCCTCCCGGAAGT[A/G]ACCCTGCCGGGCCAGGAGCCGGGAA
Phenotype
MIM: 610032
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNPO3 PubMed Links

Gene Details

Gene
TNPO3
Gene Name
transportin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191028.2 Intron NP_001177957.2
NM_012470.3 Intron NP_036602.1
XM_011515989.2 Intron XP_011514291.1
Gene
TPI1P2
Gene Name
triosephosphate isomerase 1 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details