Product Details

SNP ID
rs2722372
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:37850665 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACCAAGCCTGGTGTGGACCTTGCA[A/G]AGCAATGCAACCTTTATTCAGAAAA
Phenotype
MIM: 607421
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NME8 PubMed Links

Gene Details

Gene
NME8
Gene Name
NME/NM23 family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016616.4 500 Missense Mutation AAA,AGA K43R NP_057700.3

View Full Product Details