Product Details

SNP ID
rs12457278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5893150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAACTAACACTCTGCCATAATAG[C/T]TTTCTGACCCCTTATTAAGGTGCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM200C PubMed Links

Gene Details

Gene
TMEM200C
Gene Name
transmembrane protein 200C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080209.1 4955 Intron NP_001073678.1
XM_005258130.4 4955 Intron XP_005258187.1
XM_011525729.2 4955 UTR 5 XP_011524031.1

View Full Product Details