Product Details

SNP ID
rs905140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:6375224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTTACCAAGAGCAGGCCACCT[A/G]CCTTTTTCTGTACCCTAAAGAAATC
Phenotype
MIM: 612382
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED10 PubMed Links

Gene Details

Gene
MED10
Gene Name
mediator complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032286.2 Intron NP_115662.2

View Full Product Details